missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-1-chloroethane, 98%

Product Code. 11361427
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11361427 2 g 2g
11371427 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361427 Supplier Thermo Scientific Alfa Aesar Supplier No. L01910.04
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 593-96-4 | C2H4BrCl | 143.408 g/mol

1-Bromo-1-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-1-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigan.

Solubility
Difficult in mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-96-4
Molecular Formula C2H4BrCl
Molecular Weight (g/mol) 143.408
MDL Number MFCD00018842
InChI Key QMSVNDSDEZTYAS-UHFFFAOYSA-N
Synonym ch3chclbr, ethane, 1-bromo-1-chloro, bromochloroethane, ethane, bromochloro, bromochloroethan, chlorobromoethane, bromo-chloroethane, 1-bromochoroethane, bromo chloro ethane, acmc-20aoxm
PubChem CID 11654
IUPAC Name 1-bromo-1-chloroethane
SMILES CC(Cl)Br

Specifications

Density 1.667
Boiling Point 82°C to 83°C
Refractive Index 1.466
Quantity 2 g
UN Number UN2810
Beilstein 1730843
Solubility Information Difficult in mix.
Formula Weight 143.42
Percent Purity 98%
Chemical Name or Material 1-Bromo-1-chloroethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.