missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-1-chloroethane, 98%

Product Code. 11371427
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11371427

Brand: Thermo Scientific Alfa Aesar L01910.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 593-96-4 | C2H4BrCl | 143.408 g/mol

1-Bromo-1-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-1-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigan.

Solubility
Difficult in mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-96-4
Molecular Formula C2H4BrCl
Molecular Weight (g/mol) 143.408
MDL Number MFCD00018842
InChI Key QMSVNDSDEZTYAS-UHFFFAOYSA-N
Synonym ch3chclbr, ethane, 1-bromo-1-chloro, bromochloroethane, ethane, bromochloro, bromochloroethan, chlorobromoethane, bromo-chloroethane, 1-bromochoroethane, bromo chloro ethane, acmc-20aoxm
PubChem CID 11654
IUPAC Name 1-bromo-1-chloroethane
SMILES CC(Cl)Br

Specifications

Density 1.667
Boiling Point 82°C to 83°C
Refractive Index 1.466
Quantity 10 g
UN Number UN2810
Beilstein 1730843
Solubility Information Difficult in mix.
Formula Weight 143.42
Percent Purity 98%
Chemical Name or Material 1-Bromo-1-chloroethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.