missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-pyrrole-2-boronic acid pinacol ester, 97%

Product Code. 15411259
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411259 5 g 5g
15421259 250 mg 250mg
15401259 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411259 Supplier Thermo Scientific Alfa Aesar Supplier No. H64171.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1072944-98-9 | C15H24BNO4 | 293.17 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072944-98-9
Molecular Formula C15H24BNO4
Molecular Weight (g/mol) 293.17
MDL Number MFCD11504960
InChI Key DZQURIUJAGTAFW-UHFFFAOYSA-N
Synonym tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, 1-boc-pyrrole-2-boronic acid, pinacol ester, 1-boc-pyrrole-2-boronic acid pinacol ester, t-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole-1-carboxylate, pubchem23523, 1-boc-pyrrole-2-boronic acid,pinacol ester, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid pinacol ester, 1-boc-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, tert-butyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate
PubChem CID 46739108
IUPAC Name tert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole-1-carboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C(=O)OC(C)(C)C

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 293.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Boc-pyrrole-2-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.