missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-piperidine, 98%, Thermo Scientific™

Product Code. 11338446
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338446 1 g 1g
11348446 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338446 Supplier Thermo Scientific Alfa Aesar Supplier No. L17476.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75844-69-8
Molecular Formula C10H19NO2
Molecular Weight (g/mol) 185.267
MDL Number MFCD02093935
InChI Key RQCNHUCCQJMSRG-UHFFFAOYSA-N
Synonym 1-boc-piperidine, n-boc-piperidine, 1-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 1,1-dimethylethyl ester, piperidine-1-carboxylic acid tert-butyl ester, tert-butyl piperidinecarboxylate, boc-piperidine, n-boc piperidine, pubchem7258, n-boc tetrahydropyridine
PubChem CID 7010304
IUPAC Name tert-butyl piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCCC1

Specifications

Density 0.965
Boiling Point 130°C to 140°C (15 mmHg)
Flash Point 86°C (187°F)
Refractive Index 1.4535
Quantity 1 g
Beilstein 3604898
Formula Weight 185.27
Percent Purity 98%
Chemical Name or Material 1-Boc-piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.