missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-piperidine-4-carboxaldehyde, 97%

Product Code. 15449577
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449577 5 g 5g
15459577 250 mg 250mg
15439577 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15449577 Supplier Thermo Scientific Alfa Aesar Supplier No. H52813.06
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 137076-22-3 | C11H19NO3 | 213.28 g/mol

1-Boc-piperidine-4-carboxaldehyde is used as reactant for synthesis of Pim-1 inhibitors, selective GPR119 agonists for type II diabetes, M-tropic (R5) HIV-1 replication inhibitors, HDAC inhibitors and selective 5-HT6 antagonists. It is also used as reactant for three-component vinylogous Mannich reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-piperidine-4-carboxaldehyde is used as reactant for synthesis of Pim-1 inhibitors, selective GPR119 agonists for type II diabetes, M-tropic (R5) HIV-1 replication inhibitors, HDAC inhibitors and selective 5-HT6 antagonists. It is also used as reactant for three-component vinylogous Mannich reactions

Solubility
Soluble in methanol.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137076-22-3
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.28
MDL Number MFCD02179019
InChI Key JYUQEWCJWDGCRX-UHFFFAOYSA-N
Synonym 1-boc-4-piperidinecarboxaldehyde, 1-boc-4-formylpiperidine, 1-tert-butoxycarbonyl-4-piperidinecarboxaldehyde, 1-boc-piperidine-4-carboxaldehyde, n-boc-4-formylpiperidine, n-boc-4-piperidinecarboxaldehyde, 1-tert-butoxycarbonyl-4-formylpiperidine, 4-formyl-piperidine-1-carboxylic acid tert-butyl ester, n-boc-piperidine-4-carbaldehyde, 4-formylpiperidine-1-carboxylic acid tert-butyl ester
PubChem CID 1514430
IUPAC Name tert-butyl 4-formylpiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C=O

Specifications

Quantity 5 g
Sensitivity Air sensitive
Solubility Information Soluble in methanol.
Formula Weight 213.28
Percent Purity 97%
Chemical Name or Material 1-Boc-piperidine-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.