missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-piperidine-3-carboxaldehyde, 97%

Product Code. 15419327
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15419327 250 mg 250mg
15499317 1 g 1g
15409327 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15419327

Brand: Thermo Scientific Alfa Aesar H52571.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 118156-93-7 | C11H19NO3 | 213.277 g/mol

1-Boc-piperidine-3-carboxaldehyde is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-piperidine-3-carboxaldehyde is used as an organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118156-93-7
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.277
MDL Number MFCD02179020
InChI Key CTVHINDANRPFIL-UHFFFAOYSA-N
Synonym 1-boc-3-piperidinecarboxaldehyde, boc-3-piperidinecarboxaldehyde, 3-formylpiperidine-1-carboxylic acid tert-butyl ester, n-boc-3-formylpiperidine, 1-boc-3-formylpiperidine, 1-boc-piperidine-3-carboxaldehyde, 3-formyl-n-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester, n-boc-3-piperidine carboxyaldehyde, 1-n-boc-3-formylpiperidine
PubChem CID 9964448
IUPAC Name tert-butyl 3-formylpiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C=O

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 213.28
Percent Purity 97%
Chemical Name or Material 1-Boc-piperidine-3-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.