missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-isonipecotic acid ethyl ester, 97+%

Product Code. 11421020
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11421020 25 g 25g
11411020 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11421020

Brand: Thermo Scientific Alfa Aesar B25393.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 142851-03-4 | C13H23NO4 | 257.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142851-03-4
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD01763998
InChI Key MYHJCTUTPIKNAT-UHFFFAOYSA-N
Synonym ethyl n-boc-piperidine-4-carboxylate, 1-tert-butyl 4-ethyl piperidine-1,4-dicarboxylate, ethyl n-boc-4-piperidinecarboxylate, n-boc-4-carbethoxy piperidine, 1,4-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 4-ethyl ester, ethyl n-boc-isonipecotate, ethyl 1-boc-piperidine-4-carboxylate, dimethylethyl piperidinedicarboxylic acid ethyl ester, 1-boc-piperidine-4-carboxylic acid ethyl ester, 1-boc-isonipecotic acid ethyl ester
PubChem CID 2758812
IUPAC Name 1-O-tert-butyl 4-O-ethyl piperidine-1,4-dicarboxylate
SMILES CCOC(=O)C1CCN(CC1)C(=O)OC(C)(C)C

Specifications

Density 1.046
Boiling Point 120°C to 135°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.458
Quantity 25 g
Formula Weight 257.33
Percent Purity ≥97%
Chemical Name or Material 1-Boc-isonipecotic acid ethyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.