missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indoline-7-carboxylic acid, 97%

Product Code. 15490829
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15490829

Brand: Thermo Scientific Alfa Aesar H63883.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143262-20-8 | C14H17NO4 | 263.293 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143262-20-8
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD04973983
InChI Key SUAMIYWLXFROHE-UHFFFAOYSA-N
Synonym n-boc-indoline-7-carboxylic acid, 1-tert-butoxycarbonyl indoline-7-carboxylic acid, 2,3-dihydro-indole-1,7-dicarboxylic acid 1-tert-butyl ester, 1-tert-butoxycarbonyl-2,3-dihydroindole-7-carboxylic acid, n-boc-indoline-7-carboxylicacid, acmc-1c2ys, 1-tert-butoxycarbonyl-7-indolinecarboxylic acid, 1h-indole-1,7-dicarboxylic acid,2,3-dihydro-,1-1,1-dimethylethyl ester, 11bs-2,6-bis 3,5-bis trifluoromethyl phenyl-4-hydroxy-dinaphtho 2,1-d:1 inverted exclamation marka,2 inverted exclamation marka-f-1,3,2-dioxaphosphepin 4-oxide
PubChem CID 14977845
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-7-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC2=CC=CC(=C21)C(=O)O

Specifications

Melting Point 182°C (decomposition)
Color Yellow
Quantity 5 g
UN Number UN3077
Sensitivity Light sensitive
Formula Weight 263.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-indoline-7-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.