missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indoline-5-boronic acid pinacol ester, 97%

Product Code. 15495016
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495016 250 mg 250mg
15485758 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495016 Supplier Thermo Scientific Alfa Aesar Supplier No. H59436.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 837392-67-3 | C19H28BNO4 | 345.246 g/mol

1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 837392-67-3
Molecular Formula C19H28BNO4
Molecular Weight (g/mol) 345.246
MDL Number MFCD12408237
InChI Key OOORQXGLIKPNDK-UHFFFAOYSA-N
Synonym 1-boc-indoline-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indoline-1-carboxylate, 1,1-dimethylethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, 1h-indole-1-carboxylic acid, 2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroindole-1-carboxylate, 1-n-boc-5-bpin-indoline, amtb776, 1-tert-butoxycarbonyl-5-indolineboronic acid pinacol ester, 1-tert-butoxycarbonyl indolin-5-yl boronic acid pinacol ester
PubChem CID 18451469
IUPAC Name tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindole-1-carboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(CC3)C(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Solubility Information Soluble in dichloromethane,ethyl acetate and methanol.
Formula Weight 345.25
Percent Purity 97%
Chemical Name or Material 1-Boc-indoline-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.