missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indole, 97%

Product Code. 11368546
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368546 5 g 5g
11358546 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368546 Supplier Thermo Scientific Alfa Aesar Supplier No. L17601.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75400-67-8 | C13H15NO2 | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75400-67-8
Molecular Formula C13H15NO2
Molecular Weight (g/mol) 217.27
MDL Number MFCD02093939
InChI Key OWPIFQXNMLDXKW-UHFFFAOYSA-N
Synonym 1-boc-indole, tert-butyl 1h-indole-1-carboxylate, n-boc-indole, tert-butyl 1-indolecarboxylate, 1h-indole-1-carboxylic acid, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl indole, n-boc indole, pubchem7228, acmc-1bjgd, 1-tert-butoxycarbonylindole
PubChem CID 3532980
SMILES CC(C)(C)OC(=O)N1C=CC2=CC=CC=C12

Specifications

Density 1.07
Boiling Point 113°C to 114°C (0.2 mmHg)
Refractive Index 1.543
Quantity 5 g
Beilstein 4674239
IUPAC Name tert-butyl indole-1-carboxylate
Formula Weight 217.27
Percent Purity 97%
Chemical Name or Material 1-Boc-indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.