missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indole-2-boronic acid, 95%

Product Code. 11394865
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11394865

Brand: Thermo Scientific Alfa Aesar L18009.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 213318-44-6 | C13H16BNO4 | 261.084 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213318-44-6
Molecular Formula C13H16BNO4
Molecular Weight (g/mol) 261.084
MDL Number MFCD02093045
InChI Key SVIBPSNFXYUOFT-UHFFFAOYSA-N
Synonym n-boc-indole-2-boronic acid, 1-boc-indole-2-boronic acid, 1-tert-butoxycarbonyl indole-2-boronic acid, 1-tert-butoxycarbonyl-1h-indol-2-ylboronic acid, 1-tert-butoxycarbonyl-1h-indol-2-yl boronic acid, 1-n-boc-indole-2-boronic acid, 1-tert-butoxycarbonyl-1h-indol-2-yl-2-boronic acid, 1-tert-butoxycarbonyl indol-2-ylboronic acid, 1-boc-2-indoleboronic acid, n-boc-indol-2-yl boronic acid
PubChem CID 2773302
IUPAC Name [1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
SMILES B(C1=CC2=CC=CC=C2N1C(=O)OC(C)(C)C)(O)O

Specifications

Melting Point ∽85°C (decomposition)
Quantity 1 g
Beilstein 8148683
Formula Weight 261.08
Percent Purity 95%
Chemical Name or Material 1-Boc-indole-2-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.