missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-hexahydro-1,4-diazepine, TRC

Product Code. 30341963
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30341963 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341963 Supplier Toronto Research Chemicals Supplier No. TRCB6561752G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Boc-hexahydro-1,4-diazepine
CAS 112275-50-0
Molecular Formula C10H20N2O2
Quantity 2 g
Synonym Hexahydro-1H-1,4-Diazepine-1-carboxylic Acid 1,1-Dimethylethyl Ester, 1,1-Dimethylethyl 1,4-Diazepane-1-carboxylate, 1-(tert-Butoxycarbonyl)-1,4-diazacycloheptane, 1-(tert-Butoxycarbonyl)homopiperazine, 1-Boc-4-homopiperazine, 1-Boc-homopiperazine, 1-Homopiperazinecarboxylic Acid tert-Butyl Ester, 1-tert-Butoxycarbonyl[1,4]diazepane, 1-tert-Butoxycarbonylhexahydro-1,4-diazepane, 1-tert-Butoxycarbonylhexahydro-1,4-diazepine, BOC-homopiperazine, N-(tert-Butoxycarbonyl)homopiperazine, N-(tert-Butyloxycarbonyl)homopiperazine, [1,4]Diazepane-1-carboxylic Acid tert-Butyl Ester, tert-Butyl 1,4-Diazepane-1-carboxylate, tert-Butyl 1-homopiperazinecarboxylate, tert-Butyl Hexahydro-1H-1,4-diazepine-1-carboxylate
InChI Formula InChI=1S/C10H20N2O2/c1-10(2,3)14-9(13)12-7-4-5-11-6-8-12/h11H,4-8H2,1-3H3
SMILES CC(C)(C)OC(=O)N1CCCNCC1
IUPAC Name tert-butyl 1,4-diazepane-1-carboxylate
Molecular Weight (g/mol) 200.28
Formula Weight 200.1525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.