missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-hexahydro-1,4-diazepine, 98%

Product Code. 10604945 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10604945 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10604945 Supplier Thermo Scientific Acros Supplier No. 399530050
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 112275-50-0 | C10H20N2O2 | 200.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 112275-50-0
Boiling Point 92.0°C to 96.0°C (0.5mmHg)
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Molecular Formula C10H20N2O2
Packaging Glass Bottle
MDL Number MFCD00276987
Synonym 1-boc-homopiperazine, 1-boc-hexahydro-1,4-diazepine, 1,4 diazepane-1-carboxylic acid tert-butyl ester, 1-tert-butoxycarbonyl homopiperazine, 1-boc-1,4-diazepane, n-boc-homopiperazine, boc-hopiz, boc-homopiperazine, n-tert-butoxycarbonyl homopiperazine, tert-butyl 1-homopiperazinecarboxylate
InChI Key WDPWEXWMQDRXAL-UHFFFAOYSA-N
Density 1.0290g/mL
SMILES CC(C)(C)OC(=O)N1CCCNCC1
IUPAC Name tert-butyl 1,4-diazepane-1-carboxylate
Molecular Weight (g/mol) 200.28
Quantity 5 g
Flash Point >110°C
PubChem CID 2756058
Formula Weight 200.28
Percent Purity 98%
Specific Gravity 1.029
Physical Form Liquid
Chemical Name or Material 1-BOC-Hexahydro-1, 4-diazepine
Show More Show Less
compliance-icons
Product Identifier
  • 1-BOC-hexahydro-1,4-diazepine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.