missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-azetidine-3-methanol, 96%, Thermo Scientific Chemicals

Product Code. 15289726 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15289726 1 g 1g
15299726 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15289726

Brand: Thermo Scientific Acros 452090010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142253-56-3
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD06656141
InChI Key HXRDRJKAEYHOBB-UHFFFAOYSA-N
PubChem CID 10583745
IUPAC Name tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CO)C1

Specifications

Melting Point 50°C to 53°C
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Synonym tert-butyl 3-hydroxymethyl azetidine-1-carboxylate, 1-boc-azetidine-3-yl-methanol, 1-boc-azetidine-3-yl methanol, 1-boc-azetidine-3-ylmethanol, 1-boc-3-hydroxymethyl azetidine, 1-boc-3-azetidinemethanol, 1-tert-butoxycarbonyl-3-azetidinemethanol, 1-azetidinecarboxylic acid, 3-hydroxymethyl-, 1,1-dimethylethyl ester, tert-butyl3-hydroxymethyl azetidine-1-carboxylate, 3-hydroxymethyl-azetidine-1-carboxylic acid tert-butyl ester
Quantity 1 g
Flash Point 117°C
Formula Weight 187.24
Percent Purity 96%
Chemical Name or Material 1-BOC-azetidine-3-methanol
compliance-icons
Product Identifier
  • 1-Boc-azetidine-3-methanol, 96%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.