missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-Azetidine-3-carboxylic acid, 98%, Thermo Scientific™

Product Code. 11946731 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 11946731

Brand: Thermo Scientific Acros 442065000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 105°C to 107°C
CAS 142253-55-2
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C9H14NO4
MDL Number MFCD01860897
Synonym 1-boc-azetidine-3-carboxylic acid, 1-n-boc-3-azetidinecarboxylic acid, n-boc-azetidine-3-carboxylic acid, boc-azetidine-3-carboxylic acid, 1-tert-butoxycarbonyl azetidine-3-carboxylic acid, 1-tert-butoxy carbonyl azetidine-3-carboxylic acid, 1-n-boc-azetidine-3-carboxylic acid, boc-aze 3-oh, azetidine-1,3-dicarboxylic acid mono-tert-butyl ester, n-boc-3-azetidine carboxylic acid
InChI Key NCADHSLPNSTDMJ-UHFFFAOYSA-M
SMILES CC(C)(C)OC(=O)N1CC(C1)C([O-])=O
IUPAC Name 1-[(tert-butoxy)carbonyl]azetidine-3-carboxylate
Molecular Weight (g/mol) 200.22
Quantity 500 mg
PubChem CID 2755981
Formula Weight 201.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-BOC-Azetidine-3-carboxylic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.