missing translation for 'onlineSavingsMsg'
Learn More

1-(Boc-amino)cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 15458588
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15458588

Brand: Thermo Scientific Alfa Aesar H61992.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88950-64-5
Molecular Formula C9H15NO4
Molecular Weight (g/mol) 201.22
MDL Number MFCD00083257
InChI Key DSKCOVBHIFAJRI-UHFFFAOYSA-N
Synonym 1-boc-amino cyclopropanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropanecarboxylic acid, n-boc-1-aminocyclopropanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopropanecarboxylic acid, boc-acpc-oh, 1-tert-butoxy carbonyl amino cyclopropane-1-carboxylic acid, 1-boc amino cyclopanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 1-1,1-dimethylethoxy carbonyl amino
PubChem CID 688396
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopropane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CC1)C(O)=O

Specifications

Melting Point 118°C (decomposition)
Beilstein 4430628
Quantity 250 mg
Formula Weight 201.22
Percent Purity 98%
Chemical Name or Material 1-(Boc-amino)cyclopropanecarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.