missing translation for 'onlineSavingsMsg'
Learn More

1-(Boc-amino)cyclobutanecarboxylic acid, 97%, Thermo Scientific™

Product Code. 15459338
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459338 250 mg 250mg
15439338 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459338 Supplier Thermo Scientific Alfa Aesar Supplier No. H62888.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120728-10-1
Molecular Formula C10H16NO4
Molecular Weight (g/mol) 214.24
MDL Number MFCD02682623
InChI Key ROVVUKFHORPDSM-UHFFFAOYSA-M
Synonym n-boc-1-aminocyclobutanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 1-boc-amino cyclobutanecarboxylic acid, boc-cyclovaline, boc-acbc-oh, 1-n-boc-amino-cyclobutane carboxylic acid, 1-tert-butoxycarbonylamino cyclobutanecarboxylic acid, boc-1-aminocyclobutane-1-carboxylic acid, n-boc-1-aminocyclobutane carboxylic acid, boc-1-aminocyclobutanecarboxylic acid
PubChem CID 1512646
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCC1)C([O-])=O

Specifications

Melting Point 129°C to 133°C
Beilstein 7994003
Quantity 250 mg
Formula Weight 215.24
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Boc-amino)cyclobutanecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.