missing translation for 'onlineSavingsMsg'
Learn More

(1-Boc-4-piperidinyloxy)acetic acid, 95%, Thermo Scientific Chemicals

Product Code. 15474777
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474777 1 g 1g
15484777 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474777 Supplier Thermo Scientific Alfa Aesar Supplier No. H32102.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161948-70-5
Molecular Formula C12H21NO5
Molecular Weight (g/mol) 259.302
MDL Number MFCD02178987
InChI Key YHUAHIMRWSVXCN-UHFFFAOYSA-N
Synonym n-boc-4-carboxymethoxypiperidine, n-boc-4-carboxymethoxy-piperidine, 4-carboxymethoxy-piperidine-1-carboxylic acid tert-butyl ester, 1-boc-4-piperidinyloxy acetic acid, 2-1-tert-butoxy carbonyl piperidin-4-yl oxy acetic acid, 1-tert-butoxycarbonyl piperidin-4-yl oxy acetic acid, 1-boc-4-carboxymethoxy-piperidine, 2-1-tert-butoxycarbonyl piperidin-4-yloxy acetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yloxy aceticacid, 2-1-tert-butoxycarbonyl-4-piperidyl oxy acetic acid
PubChem CID 1502082
IUPAC Name 2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxyacetic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OCC(=O)O

Specifications

Melting Point 105°C to 107°C
Quantity 1 g
Formula Weight 259.3
Percent Purity 95%
Chemical Name or Material (1-Boc-4-piperidinyloxy)acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.