missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-4-piperidineacetic acid, 97%

Product Code. 15401418
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401418 250 mg 250mg
15491408 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15401418 Supplier Alfa Aesar™ Supplier No. H55231.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 157688-46-5
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD00800239
InChI Key ZXFLMSIMHISJFV-UHFFFAOYSA-N
Synonym 1-boc-4-piperidylacetic acid, 1-boc-piperidin-4-ylacetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl acetic acid, n-boc-4-piperidineacetic acid, 1-boc-4-piperidineacetic acid, 2-1-tert-butoxy carbonyl piperidin-4-yl acetic acid, 1-tert-butoxycarbonyl piperidin-4-yl acetic acid, boc-cmp, boc-cmp-oh
PubChem CID 1512539
IUPAC Name 2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]acetic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)CC(=O)O

Specifications

Quantity 250 mg
Beilstein 7028968
Formula Weight 243.3
Percent Purity 97%
Chemical Name or Material 1-Boc-4-piperidineacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.