missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-4-methylaminopiperidine, 95%, Thermo Scientific™

Product Code. 15274398 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15274398 5 g 5g
15264398 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15274398 Supplier Thermo Scientific Acros Supplier No. 454800050
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 147539-41-1
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.31
InChI Key CZYUGTLMFHDODF-UHFFFAOYSA-N
Synonym 1-boc-4-methylaminopiperidine, tert-butyl 4-methylamino piperidine-1-carboxylate, 1-boc-4-methylamino piperidine, 1-boc-4-methylaminopieridine, n-boc-4-methylaminopieridine, n-boc-4-methylaminopiperidine, 4-methylamino-piperidine-1-carboxylic acid tert-butyl ester, 4-methylamino piperidine, n1-boc protected, n-boc-4-n-methylamino piperidine, 4-n-methylamino-1-boc-piperidine
PubChem CID 15380702
IUPAC Name tert-butyl 4-(methylamino)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)NC

Specifications

Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 214.31
Percent Purity 95%
Chemical Name or Material 1-BOC-4-methylaminopiperidine
compliance-icons
Product Identifier
  • 1-Boc-4-methylaminopiperidine, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.