missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid, 96%

Product Code. 11414510
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414510 250 mg 250mg
11424510 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414510 Supplier Thermo Scientific Alfa Aesar Supplier No. H50086.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1210863-93-6 | C12H19NO6 | 273.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1210863-93-6
Molecular Formula C12H19NO6
Molecular Weight (g/mol) 273.29
MDL Number MFCD07781134
InChI Key OHQXZMNJROMWMW-UHFFFAOYNA-N
Synonym 4-acetoxy-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, 1-boc-4-methoxycarbonyl pyrrolidine-3-carboxylic acid, pyrrolidine-1,3,4-tricarboxylic acid 1-tert-butyl 3-methyl diester, 4-acetyloxy-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid
PubChem CID 45925717
IUPAC Name 4-acetyloxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid
SMILES CC(=O)OC1CN(CC1C(=O)O)C(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 273.29
Percent Purity 96%
Chemical Name or Material 1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.