missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-4-(Aminomethyl)piperidine, 97%

Product Code. 11936371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11936371 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11936371 Supplier Thermo Scientific Acros Supplier No. 436610050
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 144222-22-0 | C11H23N2O2 | 215.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 144222-22-0
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0130g/mL
Boiling Point 237°C to 238°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C11H23N2O2
MDL Number MFCD01076207
Quantity 5 g
Specific Gravity 1.013
Synonym 1-boc-4-aminomethyl piperidine, tert-butyl 4-aminomethyl piperidine-1-carboxylate, 1-boc-4-aminomethylpiperidine, 1-boc-4-aminomethyl-piperidine, 4-aminomethyl-1-n-boc-piperidine, boc-4-aminomethyl-piperidine, 4-aminomethyl-1-n-t-butoxycarbonyl piperidine, 4-aminomethyl-1-boc-piperidine, 1-n-boc-4-aminomethyl piperidine, 4-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester
InChI Key KLKBCNDBOVRQIJ-UHFFFAOYSA-O
SMILES CC(C)(C)OC(=O)N1CCC(C[NH3+])CC1
IUPAC Name {1-[(tert-butoxy)carbonyl]piperidin-4-yl}methanaminium
Molecular Weight (g/mol) 215.32
PubChem CID 736817
Formula Weight 214.31
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 1-BOC-4-(Aminomethyl)piperidine
Show More Show Less
compliance-icons
Product Identifier
  • 1-BOC-4-(Aminomethyl)piperidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.