missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(4-chloro-2-nitrophenyl)piperazine, 97%, Thermo Scientific™

Product Code. 15428396
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874814-97-8
Molecular Formula C15H20ClN3O4
Molecular Weight (g/mol) 341.792
InChI Key OJIDLANQYRRUCV-UHFFFAOYSA-N
Synonym tert-butyl 4-4-chloro-2-nitrophenyl piperazine-1-carboxylate
PubChem CID 69042278
IUPAC Name tert-butyl 4-(4-chloro-2-nitrophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]

Specifications

Quantity 250 mg
Beilstein 18477062
Formula Weight 341.79
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(4-chloro-2-nitrophenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'