missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(3-hydroxypropyl)piperazine, 97%, Thermo Scientific Chemicals

Product Code. 15421848
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132710-90-8
Molecular Formula C12H24N2O3
Molecular Weight (g/mol) 244.335
MDL Number MFCD06798090
InChI Key LRYRQGKGCIUVON-UHFFFAOYSA-N
Synonym 1-boc-4-3-hydroxypropyl piperazine, tert-butyl 4-3-hydroxypropyl piperazine-1-carboxylate, 4-2-hydroxypropyl-1-piperazinecarboxylic acid, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-3-hydroxypropyl piperazine, 1-piperazinecarboxylicacid, 4-3-hydroxypropyl-, 1,1-dimethylethyl ester, acmc-20dtb2, t-butyl 4-3-hydroxypropyl piperazine-1-carboxylate, 1-3-hydroxypropyl-4-tert-butyloxycarbonylpiperazine
PubChem CID 16217800
IUPAC Name tert-butyl 4-(3-hydroxypropyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CCCO

Specifications

Melting Point 73°C to 77°C
Quantity 1 g
Formula Weight 244.33
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(3-hydroxypropyl)piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'