missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-methoxycarbonylphenyl)piperazine, 97%, Thermo Scientific™

£22.70 - £216.00

Chemical Identifiers

CAS 870703-74-5
Molecular Formula C17H24N2O4
Molecular Weight (g/mol) 320.39
MDL Number MFCD07369760
InChI Key JFZGCXORLWXLGH-UHFFFAOYSA-N
Synonym 1-boc-4-2-methoxycarbonylphenyl piperazine, tert-butyl 4-2-methoxycarbonyl phenyl piperazine-1-carboxylate, methyl 2-boc-piperazin-1-yl benzoate, 1-boc-4-2-methoxycarbonylphenyl pipera, 1-boc-4-2-methoxycarbonyl phenyl-piperazine
PubChem CID 16217924
IUPAC Name tert-butyl 4-(2-methoxycarbonylphenyl)piperazine-1-carboxylate
SMILES COC(=O)C1=CC=CC=C1N1CCN(CC1)C(=O)OC(C)(C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15482016
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55302.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15441518
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55302.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15451518
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55302.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

870703-74-5
320.39
JFZGCXORLWXLGH-UHFFFAOYSA-N
16217924
COC(=O)C1=CC=CC=C1N1CCN(CC1)C(=O)OC(C)(C)C
C17H24N2O4
MFCD07369760
1-boc-4-2-methoxycarbonylphenyl piperazine, tert-butyl 4-2-methoxycarbonyl phenyl piperazine-1-carboxylate, methyl 2-boc-piperazin-1-yl benzoate, 1-boc-4-2-methoxycarbonylphenyl pipera, 1-boc-4-2-methoxycarbonyl phenyl-piperazine
tert-butyl 4-(2-methoxycarbonylphenyl)piperazine-1-carboxylate

Specifications

870703-74-5
>110°C (230°F)
1.532
UN2810
JFZGCXORLWXLGH-UHFFFAOYSA-N
tert-butyl 4-(2-methoxycarbonylphenyl)piperazine-1-carboxylate
16217924
97%
1-Boc-4-(2-methoxycarbonylphenyl)piperazine
Yellow to Orange
C17H24N2O4
MFCD07369760
1-boc-4-2-methoxycarbonylphenyl piperazine, tert-butyl 4-2-methoxycarbonyl phenyl piperazine-1-carboxylate, methyl 2-boc-piperazin-1-yl benzoate, 1-boc-4-2-methoxycarbonylphenyl pipera, 1-boc-4-2-methoxycarbonyl phenyl-piperazine
COC(=O)C1=CC=CC=C1N1CCN(CC1)C(=O)OC(C)(C)C
320.39
320.39
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications