missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-Fluoro-6-nitrophenyl)piperazine, 97%, Thermo Scientific™

Catalog Number 15418446
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1233958-36-5
C15H20FN3O4
325.34
MFCD14155993
GWHUVEIGDXIWOY-UHFFFAOYSA-N
tert-butyl 4-2-fluoro-6-nitrophenyl piperazine-1-carboxylate, 1-boc-4-2-fluoro-6-nitrophenyl piperazine
53255503
tert-butyl 4-(2-fluoro-6-nitrophenyl)piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=CC=C2F)[N+](=O)[O-]
This item is not returnable. View return policy
1233958-36-5
C15H20FN3O4
325.34
MFCD14155993
GWHUVEIGDXIWOY-UHFFFAOYSA-N
tert-butyl 4-2-fluoro-6-nitrophenyl piperazine-1-carboxylate, 1-boc-4-2-fluoro-6-nitrophenyl piperazine
53255503
tert-butyl 4-(2-fluoro-6-nitrophenyl)piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=CC=C2F)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 19933459
Formula Weight 325.34
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(2-Fluoro-6-nitrophenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'