missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-Fluoro-4-nitrophenyl)piperazine, 97%, Thermo Scientific™

Product Code. 15458256
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458256 250 mg 250mg
15429988 1 g 1g
15439988 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458256 Supplier Thermo Scientific Alfa Aesar Supplier No. H63340.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154590-34-8
Molecular Formula C15H20FN3O4
Molecular Weight (g/mol) 325.34
MDL Number MFCD08443959
InChI Key YOLVWCABLVHASL-UHFFFAOYSA-N
PubChem CID 16203508
IUPAC Name tert-butyl 4-(2-fluoro-4-nitrophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F

Specifications

Quantity 250 mg
Synonym tert-butyl 4-2-fluoro-4-nitrophenyl piperazine-1-carboxylate, tert-butyl 4-2-fluoro-4-nitrophenyl tetrahydro-1 2h-pyrazinecarboxylate, butylfluoronitrophenyltetrahydropyrazinecarboxylate, 1-boc-4-2-fluoro-4-nitrophenyl piperazine, 1-tert-butoxycarbonyl-4-2-fluoro-4-nitrophenyl piperazine, 3-fluoro-4-4-tert-butoxycarbonyl piperazin-1-yl nitrobenzene, tert-butyl4-2-fluoro-4-nitrophenyl piperazine-1-carboxylate, 4-2-fluoro-4-nitro-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-fluoro-4-nitro-phenyl piperazine-1-carboxylic acid tert-butyl ester, 4-2-fluoro-4-nitrophenyl piperazine-1-carboxylic acid tert-butyl ester
Formula Weight 325.34
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(2-Fluoro-4-nitrophenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.