missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-chloroethyl)piperazine, 97%

Product Code. 15406167
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406167 1 g 1g
15416167 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406167 Supplier Thermo Scientific Alfa Aesar Supplier No. H33349.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 208167-83-3 | C11H21ClN2O2 | 248.751 g/mol

By the direct condensation of chloroalkyl piperazine, nitrogen mustard benzaldehyde, and pyrrole anticancer activity to bel-7404 liver cancer cells was found.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
By the direct condensation of chloroalkyl piperazine, nitrogen mustard benzaldehyde, and pyrrole anticancer activity to bel-7404 liver cancer cells was found.

Solubility
Slightly Soluble (7.2 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208167-83-3
Molecular Formula C11H21ClN2O2
Molecular Weight (g/mol) 248.751
MDL Number MFCD08443993
InChI Key MYOWELLYEZMECA-UHFFFAOYSA-N
Synonym tert-butyl 4-2-chloroethyl piperazine-1-carboxylate, tert-butyl 4-2-chloroethyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-chloroethyl piperazine, tert-butyl 4-2-chloroethyl piperazinecarboxylate, 1-piperazinecarboxylic acid, 4-2-chloroethyl-, 1,1-dimethylethyl ester, 4-2-chloroethyl piperazine, n1-boc protected, 4-2-chloroethyl piperazine,n1-boc protected, tertbutylchloroethyltetrahydropyrazinecarboxylate, tert-butyl 4-2-chloroethyl tetrahydro-1 2h-pyraz
PubChem CID 22106269
IUPAC Name tert-butyl 4-(2-chloroethyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CCCl

Specifications

Melting Point 63°C to 65°C
Quantity 1 g
Solubility Information Slightly Soluble (7.2g/L) (25°C).
Formula Weight 248.76
Percent Purity 97%
Chemical Name or Material 1-Boc-4-(2-chloroethyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.