missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-3-pyrrolidinol, TRC
Click to view available options
Quantity:
2 g
Unit Size:
2g
Specifications
Specifications
| Chemical Name or Material | 1-Boc-3-pyrrolidinol |
| CAS | 103057-44-9 |
| Molecular Formula | C9 H17 N O3 |
| Quantity | 2 g |
| Synonym | 1-Pyrrolidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, (RS)-3-Hydroxypyrrolidine-1-carboxylic acid tert-butyl ester, 1,1-Dimethylethyl 3-hydroxypyrrolidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-pyrrolidinol, 1-(tert-Butyloxycarbonyl)-3-hydroxypyrrolidine, 1-Boc-3-hydroxypyrrolidine, 1-Boc-3-pyrrolidinol, 1-tert-Butoxycarbonyl-3-hydroxypyrrolidine, 3-Hydroxy-pyrrolidin-1-carboxylic acid tert-butyl ester, 3-Hydroxypyrrolidine-1-carboxylic acid tert-butyl ester, N-(tert-Butoxycarbonyl)-3-hydroxypyrrolidine, N-tert-Butyloxycarbonyl-3-pyrrolidinol, tert-Butyl 3-hydroxy-1-pyrrolidinecarboxylate |
| InChI Formula | InChI=1S/C9H17NO3/c1-9(2,3)13-8(12)10-5-4-7(11)6-10/h7,11H,4-6H2,1-3H3 |
| SMILES | CC(C)(C)OC(=O)N1CCC(O)C1 |
| IUPAC Name | tert-butyl 3-hydroxypyrrolidine-1-carboxylate |
| Molecular Weight (g/mol) | 187.24 |
| Formula Weight | 187.1208 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction