missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Boc-3-hydroxypyrrolidine, 97%

Product Code. 11442130
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442130 25 g 25g
11432130 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442130 Supplier Thermo Scientific Alfa Aesar Supplier No. H28750.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103057-44-9 | C9H17NO3 | 187.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103057-44-9
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.239
MDL Number MFCD04038535
InChI Key APCBTRDHCDOPNY-UHFFFAOYSA-N
Synonym 1-boc-3-hydroxypyrrolidine, 1-boc-3-pyrrolidinol, n-boc-3-hydroxypyrrolidine, n-boc-3-pyrrolidinol, 1-n-boc-3-hydroxypyrrolidine, 1-pyrrolidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-pyrrolidinol, 3-hydroxy-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-hydroxypyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-hydroxypyrrolidine
PubChem CID 4416939
IUPAC Name tert-butyl 3-hydroxypyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)O

Specifications

Melting Point 60°C to 62°C
Quantity 25 g
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material (+/-)-1-Boc-3-hydroxypyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.