missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-hydroxyazetidine, 97%

Product Code. 11452180
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452180 5 g 5g
15453837 250 mg 250mg
11442180 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452180 Supplier Thermo Scientific Alfa Aesar Supplier No. H28902.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 141699-55-0 | C8H15NO3 | 173.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141699-55-0
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.212
MDL Number MFCD04115305
InChI Key XRRXRQJQQKMFBC-UHFFFAOYSA-N
Synonym 1-n-boc-3-hydroxyazetidine, n-boc-3-hydroxyazetidine, 1-boc-3-hydroxyazetidine, 1-boc-3-hydroxy azetidine, 1-boc-3-azetidinol, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-azetidin-3-ol, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester
PubChem CID 2756801
IUPAC Name tert-butyl 3-hydroxyazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)O

Specifications

Melting Point 40°C to 47°C
Quantity 5 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material 1-Boc-3-hydroxyazetidine
compliance-icons
Product Identifier
  • 1-Boc-3-hydroxyazetidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-BOC-3-Hydroxyazetidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.