missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-Hydroxyazetidine, 97%, Thermo Scientific Chemicals

Product Code. 11916731 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11916731

Brand: Thermo Scientific Acros 441760050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141699-55-0
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
InChI Key XRRXRQJQQKMFBC-UHFFFAOYSA-N
Synonym 1-n-boc-3-hydroxyazetidine, n-boc-3-hydroxyazetidine, 1-boc-3-hydroxyazetidine, 1-boc-3-hydroxy azetidine, 1-boc-3-azetidinol, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-azetidin-3-ol, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester
PubChem CID 2756801
IUPAC Name tert-butyl 3-hydroxyazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)O

Specifications

Melting Point 39.0°C to 44.0°C
CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 173.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-BOC-3-Hydroxyazetidine
Show More Show Less
compliance-icons
Product Identifier
  • 1-BOC-3-Hydroxyazetidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.