missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-fluoro-4-piperidone, 95%, Thermo Scientific™

Product Code. 15466057
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466057 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15466057 Supplier Thermo Scientific Alfa Aesar Supplier No. H33244.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 211108-50-8
Melting Point 88°C to 90°C
Molecular Formula C10H16FNO3
MDL Number MFCD10566554
Quantity 250 mg
Synonym 1-boc-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-boc-3-fluoro-4-piperidone, 1-tert-butoxycarbonyl-3-fluoro-4-piperidone, 1-piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl3-fluoro-4-oxopiperidine-1-carboxylate, tert-butyl 3-fluoro-4-oxo-piperidine-1-carboxylate, n-boc-3-fluoro-4-piperidinone
InChI Key JZNWQLLPLOQGOI-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CCC(=O)C(C1)F
IUPAC Name tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate
Molecular Weight (g/mol) 217.24
PubChem CID 10560711
Formula Weight 217.24
Percent Purity 95%
Chemical Name or Material 1-Boc-3-fluoro-4-piperidone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.