missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-(bromomethyl)azetidine, 95%

Product Code. 15443958
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
Product Code. Quantity unitSize
15443958 250 mg 250mg
15453958 500 mg 500mg
2 options
This item is not returnable. View return policy

Product Code. 15443958

Brand: Thermo Scientific Alfa Aesar H57492.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 253176-93-1 | C9H16BrNO2 | 250.14 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Sparingly soluble in water at 25°C, 0.83 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253176-93-1
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.14
MDL Number MFCD16556174
InChI Key PUKCUGDJEPVLPR-UHFFFAOYSA-N
Synonym 1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate
PubChem CID 53350331
IUPAC Name tert-butyl 3-(bromomethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CBr)C1

Specifications

Quantity 250 mg
Solubility Information Sparingly soluble in water at 25°C,0.83g/L.
Formula Weight 250.13
Percent Purity 95%
Chemical Name or Material 1-Boc-3-(bromomethyl)azetidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.