missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-3-azetidinone, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 1-Boc-3-azetidinone |
| CAS | 398489-26-4 |
| Molecular Formula | C8H13NO3 |
| Quantity | 100 mg |
| Synonym | 3-Oxo-1-azetidinecarboxylic Acid 1,1-Dimethylethyl Ester, 1-Boc-3-oxo-azetidine, 1-tert-Butoxycarbonyl-3-azetidinone, 3-Oxoazetidine-1-carboxylic Acid tert-Butyl Ester, 3-Oxoazetidine-1-carboxylic Acid tert-Butyl Ester, tert-Butyl 3-Oxoazetidine-1-carboxylate |
| InChI Formula | InChI=1S/C8H13NO3/c1-8(2,3)12-7(11)9-4-6(10)5-9/h4-5H2,1-3H3 |
| SMILES | CC(C)(C)OC(=O)N1CC(=O)C1 |
| IUPAC Name | tert-butyl 3-oxoazetidine-1-carboxylate |
| Molecular Weight (g/mol) | 171.19 |
| Formula Weight | 171.0895 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction