missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-Azetidinone, 97%

Product Code. 11995431 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11995431

Brand: Thermo Scientific Acros 442040010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 398489-26-4 | C8H13NO3 | 171.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 398489-26-4
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.20
MDL Number MFCD01861741
InChI Key VMKIXWAFFVLJCK-UHFFFAOYSA-N
Synonym 1-boc-3-azetidinone, 1-boc-azetidin-3-one, 1-n-boc-3-azetidinone, n-boc-3-azetidinone, n-boc-3-oxoazetidine, 1-tert-butoxycarbonyl-3-azetidinone, tert-butyl-3-oxoazetidine-1-carboxylate, 1-boc-3-oxoazetidine, n-boc-azetidin-3-one, 3-oxo-azetidine-1-carboxylic acid tert-butyl ester
PubChem CID 1519404
IUPAC Name tert-butyl 3-oxoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50°C to 52°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 171.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-BOC-3-Azetidinone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.