missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-2-trifluoromethyl-4-piperidone, 95%

Product Code. 15412759
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412759 250 mg 250mg
15402759 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15412759 Supplier Thermo Scientific Alfa Aesar Supplier No. H66437.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1245648-32-1 | C11H16F3NO3 | 267.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1245648-32-1
Molecular Formula C11H16F3NO3
Molecular Weight (g/mol) 267.25
MDL Number MFCD07367733
InChI Key RSFRBHIRISNXIK-UHFFFAOYNA-N
PubChem CID 71607278
IUPAC Name tert-butyl 4-oxo-2-(trifluoromethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)CC1C(F)(F)F

Specifications

Quantity 250 mg
Synonym tert-butyl 4-oxo-2-trifluoromethyl piperidine-1-carboxylate, 1-boc-2-trifluoromethyl-piperidin-4-one, 1-boc-2-trifluromethylpiperidin-4-one, 1-boc-2-trifluoromethylpiper idin-4-one, t-butyl 4-oxo-2-trifluoromethyl piperidine-1-carboxylate, tert-butyl4-oxo-2-trifluoromethyl piperidine-1-carboxylate, tert-butyl 4-oxo-2-trifluoromethyl ;piperidine-1-carboxylate, 1-boc-2-trifluromethyl-piperidin-4-one;4-oxo-2-trifluoromethyl-piperidine-1-carboxylic acid tert-butyl ester
Formula Weight 267.25
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1-Boc-2-trifluoromethyl-4-piperidone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.