missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-2,6-diazaspiro[3.3]heptane hemioxalate, 97%, Thermo Scientific™

Product Code. 15448506
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448506 250 mg 250mg
15400629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448506 Supplier Thermo Scientific Alfa Aesar Supplier No. H63763.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1041026-71-4
Molecular Formula C22H38N4O8
Molecular Weight (g/mol) 486.566
MDL Number MFCD12404932
InChI Key PYRAMVPOMDHTNX-UHFFFAOYSA-N
Synonym tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate oxalate 2:1, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxylate, tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate hemioxalate, tert-butyl 2,6-diazaspiro 3,3 heptane-2-carboxylate hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid, 1,1-dimethylethyl ester, ethanedioate 2:1, oxalic acid; bis tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate, 2-boc-2,6-diazaspiro 3.3 heptaneoxalate, 2-boc-2,6-diazaspiro 3.3 heptane hemioxalate, 1-boc-2,6-diazaspiro 3.3 heptane hemioxalate
PubChem CID 50902214
IUPAC Name tert-butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate;oxalic acid
SMILES CC(C)(C)OC(=O)N1CC2(C1)CNC2.CC(C)(C)OC(=O)N1CC2(C1)CNC2.C(=O)(C(=O)O)O

Specifications

Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 441.54
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-2,6-diazaspiro[3.3]heptane hemioxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.