missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-1,2,3,6-tetrahydropyridine, 97%
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Description
1-Boc-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Boc-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
1-Boc-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications
Specifications
| Boiling Point | 78°C to 84°C (5mmHg) |
| Solubility Information | Insoluble in water. Soluble in organic solvents. |
| Density | 1.029 |
| Quantity | 1 g |
| Formula Weight | 183.25 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-Boc-1,2,3,6-tetrahydropyridine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction