missing translation for 'onlineSavingsMsg'
Learn More

1-Beta-D-Ribofuranosyl-1,2,4-triazole-3-carboxylic Acid Methyl Ester (Ribavirin Impurity H), TRC

Product Code. 30422457
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30422457 100 mg
30411178 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30422457 Supplier Toronto Research Chemicals Supplier No. TRCR415725100MG
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38934-69-9
Molecular Formula C9 H13 N3 O7
Molecular Weight (g/mol) 275.22
Synonym 1-β-D-Ribofuranosyl-1H-1,2,4-triazole-3-carboxylic Acid Methyl Ester, Methyl 1-(β-D-Ribofuranosyl)-1,2,4-triazole-3-carboxylate, NSC 360639
IUPAC Name [(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl] 1H-1,2,4-triazol-3-ylmethyl carbonate
SMILES OC[C@H]1O[C@@H](OC(=O)OCc2nc[nH]n2)[C@H](O)[C@@H]1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H13N3O7/c13-1-4-6(14)7(15)8(18-4)19-9(16)17-2-5-10-3-11-12-5/h3-4,6-8,13-15H,1-2H2,(H,10,11,12)/t4-,6-,7-,8+/m1/s1
Formula Weight 275.0753
Chemical Name or Material 1-beta-D-Ribofuranosyl-1,2,4-triazole-3-carboxylic acid methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.