missing translation for 'onlineSavingsMsg'
Learn More

1-beta-D-Arabinofuranosyluracil, MedChemExpress

Product Code. 16489186
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16489186 Supplier Medchem Express Supplier No. HYN6652
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 1-beta-D-Arabinofuranosyluracil (Uracil 1-β-D-arabinofuranoside) isolated from the Caribbean sponge Tectitethya crypta, is a methoxyadenosine derivative. 1-beta-D-Arabinofuranosyluracil has demonstrated a diverse bioactivity profile including anti-inflammatory activity, analgesic and vasodilation properties. 1-beta-D-Arabinofuranosyluracil reduces a proliferation of mouse lymphoma cells.

1-beta-D-Arabinofuranosyluracil (Uracil 1-β-D-arabinofuranoside) isolated from the Caribbean sponge Tectitethya crypta, is a methoxyadenosine derivative. 1-beta-D-Arabinofuranosyluracil has demonstrated a diverse bioactivity profile including anti-inflammatory activity, analgesic and vasodilation properties. 1-beta-D-Arabinofuranosyluracil reduces a proliferation of mouse lymphoma cells.

Specifications

Chemical Name or Material 1-beta-D-Arabinofuranosyluracil
CAS 3083-77-0
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C9H12N2O6
Quantity 100 mg
Synonym Uracil 1-β-D-arabinofuranoside
Solubility Information DMSO : ≥ 100 mg/mL (409.50 mM)
SMILES O=C1NC(C=CN1[C@H]2[C@H]([C@@H]([C@@H](CO)O2)O)O)=O
Molecular Weight (g/mol) 244.2
Formula Weight 244.2
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.