missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylpyrrole, 96%

Product Code. p-7052811
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15462338

Brand: Thermo Scientific Alfa Aesar H55936.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2051-97-0 | C11H11N | 157.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-97-0
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.22
MDL Number MFCD00963301
InChI Key FNEQHKCQXDKYEO-UHFFFAOYSA-N
Synonym n-benzylpyrrole, 1h-pyrrole, 1-phenylmethyl, 1-benzyl-1h-pyrrole, pyrrole, 1-benzyl, 1-benzylpyrrol, acmc-209fbl, 1-phenylmethyl pyrrole, 1-benzyl-1h-pyrrole #, pyrrole, 1-benzyl-8ci
PubChem CID 74922
SMILES C(N1C=CC=C1)C1=CC=CC=C1

Specifications

Color Colorless to Brown
Density 1.020 g/mL
Boiling Point 90°C (0.9 mmHg)
Refractive Index 1.5685
Quantity 1 g
UN Number UN1993
IUPAC Name 1-benzyl-1H-pyrrole
Formula Weight 157.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Benzylpyrrole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.