missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylpiperidine, 98%

Product Code. p-7047501
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11384256

Brand: Thermo Scientific Alfa Aesar L19339.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2905-56-8 | C12H18ClN | 211.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-56-8
Molecular Formula C12H18ClN
Molecular Weight (g/mol) 211.73
MDL Number MFCD00224901
InChI Key MPPIBJJDFLONMO-UHFFFAOYSA-N
Synonym piperidine, 1-phenylmethyl, benzylpiperidine, unii-7hze16210b, n-benzylpiperidine, benzyl-piperidine, n-benzyl piperidine, n-benzyl-piperidine, 1-benzyl-piperidine, acmc-20a2pl
PubChem CID 76190
SMILES [H+].[Cl-].C(N1CCCCC1)C1=CC=CC=C1

Specifications

Density 0.95
Boiling Point 120°C to 123°C (9 mmHg)
Refractive Index 1.526
Quantity 10 g
Beilstein 5639
IUPAC Name 1-benzylpiperidine
Formula Weight 175.28
Percent Purity 98%
Chemical Name or Material 1-Benzylpiperidine
compliance-icons
Product Identifier
  • 1-Benzylpiperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Benzylpiperidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.