missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Benzyloxycarbonyl-4-Boc-piperazine-2-carboxylic acid, 97%

Product Code. 11496080
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11496080

Brand: Thermo Scientific Alfa Aesar H27047.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 126937-41-5 | C18H24N2O6 | 364.398 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126937-41-5
Molecular Formula C18H24N2O6
Molecular Weight (g/mol) 364.398
MDL Number MFCD01631192
InChI Key MKXMXZZARNRMMQ-UHFFFAOYSA-N
Synonym n-4-boc-n-1-cbz-2-piperazine carboxylic acid, 4-boc-1-cbz-2-piperazinecarboxylic acid, 1-benzyloxy carbonyl-4-tert-butoxycarbonyl piperazine-2-carboxylic acid, 4-boc-1-cbz-2-piperazinecarboxylicacid, 1-benzyloxycarbonyl-4-tert-butoxycarbonyl piperazine-2-carboxylic acid, 4-boc-1-cbz-piperazine-2-carboxylic acid, 4-n-boc-1-n-cbz-piperazine-2-carboxylic acid, n-1-boc-n-4-cbz-2-piperazinecarboxylic acid t-butyl ester, +/--1-benzyloxycarbonyl-4-boc-piperazine-2-carboxylic acid
PubChem CID 2756777
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonyl]-1-phenylmethoxycarbonylpiperazine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCN(C(C1)C(=O)O)C(=O)OCC2=CC=CC=C2

Specifications

Melting Point 150°C to 152°C
Quantity 5 g
Formula Weight 364.4
Percent Purity 97%
Chemical Name or Material (+/-)-1-Benzyloxycarbonyl-4-Boc-piperazine-2-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.