missing translation for 'onlineSavingsMsg'
Learn More

1-[(Benzyloxy)carbonyl]-3-piperidinecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10386514
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10386514 25 g 25g
10486272 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10386514 Supplier Thermo Scientific Maybridge Supplier No. CC24701EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 78190-11-1
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD02931282
InChI Key FFLPIVZNYJKKDM-UHFFFAOYSA-N
Synonym 1-benzyloxy carbonyl-3-piperidinecarboxylic acid, 1-benzyloxy carbonyl piperidine-3-carboxylic acid, n-cbz-piperidine-3-carboxylic acid, 1-cbz-3-piperidinecarboxylic acid, 1-n-cbz-piperidine-3-carboxylic acid, n-cbz-3-piperidinecarboxylic acid, n-cbz-dl-nipecotic acid, 1-benzyloxycarbonyl piperidine-3-carboxylic acid, 1-n-cbz-piperidin-3-ylcarboxylic acid, 1,3-piperidinedicarboxylic acid, 1-phenylmethyl ester
PubChem CID 234339
IUPAC Name 1-phenylmethoxycarbonylpiperidine-3-carboxylic acid
SMILES C1CC(CN(C1)C(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 263.29
Percent Purity 97%
Chemical Name or Material 1-[(Benzyloxy)carbonyl]-3-piperidinecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.