missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxy-4-iodobenzene, 98+%

Product Code. 11342887
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342887 25 g 25g
11332887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342887 Supplier Thermo Scientific Alfa Aesar Supplier No. L16092.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19578-68-8 | C13H11IO | 310.134 g/mol

1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19578-68-8
Molecular Formula C13H11IO
Molecular Weight (g/mol) 310.134
MDL Number MFCD00079704
InChI Key MPWFGAWFTAZWKZ-UHFFFAOYSA-N
PubChem CID 519671
IUPAC Name 1-iodo-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)I

Specifications

Melting Point 60°C to 64°C
Boiling Point 140°C (1 mmHg)
Quantity 25 g
Beilstein 3051495
Sensitivity Light sensitive
Synonym 1-benzyloxy-4-iodobenzene, 4-benzyloxyiodobenzene, 4-iodobenzyloxybenzene, benzyl 4-iodophenyl ether, benzene, 1-iodo-4-phenylmethoxy, 1-benzyloxy-4-iodo-benzene, benzene,1-iodo-4-phenylmethoxy, acmc-20amon, 4-benzyloxy iodobenzene
Solubility Information Insoluble in water.
Formula Weight 310.14
Percent Purity ≥98%
Chemical Name or Material 1-Benzyloxy-4-iodobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.