missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylindole, 97%

Product Code. 15400099
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15400099

Brand: Thermo Scientific Alfa Aesar H63414.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3377-71-7 | C15H13N | 207.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3377-71-7
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00015884
InChI Key NJZQOCCEDXRQJM-UHFFFAOYSA-N
Synonym 1-benzyl-1h-indole, n-benzylindole, 1-benzyl-indole, 1h-indole, 1-phenylmethyl, n-benzyl indole, n-benzyl-indole, zlchem 819, phenylmethyl-1h-indole, cambridge id 5106611, 1h-indole, phenylmethyl
PubChem CID 96913
IUPAC Name 1-benzylindole
SMILES C1=CC=C(C=C1)CN2C=CC3=CC=CC=C32

Specifications

Color White to Yellow
Quantity 1 g
Beilstein 148884
Formula Weight 207.27
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Benzylindole
compliance-icons
Product Identifier
  • 1-Benzylindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Benzylindole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.