missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylindole-3-carboxylic acid, 98%, Thermo Scientific™

Product Code. p-4089901 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10008249

Brand: Thermo Scientific Acros 428260010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 27018-76-4
Molecular Formula C16H13NO2
Molecular Weight (g/mol) 251.28
MDL Number MFCD00057094
InChI Key LVYDDRHDOKXFMW-UHFFFAOYSA-N
Synonym 1-benzyl-1h-indole-3-carboxylic acid, indole-3-carboxylic acid, 1-benzyl, 1h-indole-3-carboxylic acid, 1-phenylmethyl, pubchem7313, cbmicro_033327, cambridge id 5791025, 5-22-03-00037 beilstein handbook reference, 1-benzylindol-3-carboxylic acid, n-benzylindole-3-carboxylic acid, 1h-indole-3-carboxylicacid, 1-phenylmethyl
PubChem CID 33671
IUPAC Name 1-benzylindole-3-carboxylic acid
SMILES C1=CC=C(C=C1)CN2C=C(C3=CC=CC=C32)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 251.28
Percent Purity 98%
Chemical Name or Material 1-Benzylindole-3-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.