missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylimidazole, 98+%

Product Code. 11310234
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11310234 5 g 5g
11320234 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310234 Supplier Thermo Scientific Alfa Aesar Supplier No. L00802.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4238-71-5 | C10H10N2 | 158.20 g/mol

1-Benzylimidazole acts as an inducer of various cytochrome P-450 isozymes and inhibitor of thromboxane A2 synthase. 1-Benzylimidazole has been used to prepare cyclodextrin-ionic liquid polymer (βCD-BIMOTs-TDI)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzylimidazole acts as an inducer of various cytochrome P-450 isozymes and inhibitor of thromboxane A2 synthase. 1-Benzylimidazole has been used to prepare cyclodextrin-ionic liquid polymer (βCD-BIMOTs-TDI)

Solubility
Soluble in methanol, acetone, DMSO, ethanol, dilute acids (freely soluble), and buffers of pH>6.0 (freely soluble). Insoluble in water.

Notes
Store in a cool, dry place. Keep container closed when not in use. Incompatible with Oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4238-71-5
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00005296
InChI Key KKKDZZRICRFGSD-UHFFFAOYSA-N
Synonym 1-benzyl-1h-imidazole, n-benzylimidazole, imidazole, 1-benzyl, benzylimidazole, 1h-imidazole, 1-phenylmethyl, 1-benzyl imidazole, ccris 5821, chembl14192, 1bn, 1-benzylimdazole
PubChem CID 77918
IUPAC Name 1-benzylimidazole
SMILES C(N1C=CN=C1)C1=CC=CC=C1

Specifications

Melting Point 68°C to 73°C
Boiling Point 310°C
Quantity 5 g
Beilstein 114571
Solubility Information Soluble in methanol,acetone,DMSO,ethanol,dilute acids (freely soluble),and buffers of pH>6.0 (freely soluble). Insoluble in water.
Formula Weight 158.2
Percent Purity ≥98%
Chemical Name or Material 1-Benzylimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.