missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-5-phenylbarbituric acid, 98+%

Product Code. 11347816
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347816 5 g 5g
11357816 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347816 Supplier Thermo Scientific Alfa Aesar Supplier No. L12954.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 72846-00-5 | C17H14N2O3 | 294.31 g/mol

1-Benzyl-5-phenylbarbituric acid is useful as a pharmaceutical intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72846-00-5
Molecular Formula C17H14N2O3
Molecular Weight (g/mol) 294.31
MDL Number MFCD00082750
InChI Key KCWWCWMGJOWTMY-UHFFFAOYNA-N
Synonym 1-benzyl-5-phenylbarbituric acid, 1-benzyl-5-phenylpyrimidine-2,4,6 1h,3h,5h-trione, benzyl-5-phenylbarbituric acid, 2,4,6 1h,3h,5h-pyrimidinetrione, 5-phenyl-1-phenylmethyl, 1-phenylmethyl-5-phenyl-barbituric acid, 5-phenyl-1-benzyl-1,3,5-trihydropyrimidine-2,4,6-trione, acmc-1bb85, 1-benzyl-5-phenylbarbiturate, 1-benzyl-5-phenylbarbituricacid, 1-benzyl-5-phenylbarbituric acid 5g
PubChem CID 166340
IUPAC Name 1-benzyl-5-phenyl-1,3-diazinane-2,4,6-trione
SMILES O=C1NC(=O)N(CC2=CC=CC=C2)C(=O)C1C1=CC=CC=C1

Specifications

Melting Point 163°C to 165°C
Formula Weight 294.31
Percent Purity ≥98%
Quantity 5 g
Chemical Name or Material 1-Benzyl-5-phenylbarbituric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.